1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine 99%

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Piperazin-1-yl)-3-(trifluoromethyl)pyridine, 1-(3-(Trifluoromethyl)pyridin-2-yl)piperazine

PC9120-1G PC9120-5G PC9120-10G
APOSPC9120-1GEA 40.8 EUR
APOSPC9120-1G APOSPC9120-5G APOSPC9120-10G
1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine 99%
1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine
Formula: C₁₀H₁₂F₃N₃
MW: 231,218 g/mol
Boiling Pt: 106 °C (1,5 mm Hg)
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00114705
CAS Number: 87394-63-6
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR