1,4-Bis[chloro(difluoro)methyl]benzene 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
α,α'-Dichloro-α,α,α',α'-tetrafluoro-p-xylene
PC9183-25G
PC9183-5G
PC9183-1G
APOSPC9183-25GEA
343
EUR
APOSPC9183-25G
APOSPC9183-5G
APOSPC9183-1G
1,4-Bis[chloro(difluoro)methyl]benzene 98%
1,4-Bis[chloro(difluoro)methyl]benzene
Formula:
C₈H₄Cl₂F₄ MW: 247,017 g/mol Boiling Pt: 55,5 °C (3.8 mm Hg) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320797 CAS Number: 2629-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...