4-Methyl-3-(trifluoromethyl)cinnamic acid 97%
Supplier: Apollo Scientific
PC9184-1G
PC9184-5G
APOSPC9184-1GEA
159
EUR
APOSPC9184-1G
APOSPC9184-5G
4-Methyl-3-(trifluoromethyl)cinnamic acid 97%
4-Methyl-3-(trifluoromethyl)cinnamic acid
Formula:
C₁₁H₉F₃O₂ MW: 230,183 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116034 CAS Number: 886498-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...