2-Chloro-5-(trifluoromethyl)cinnamic acid 98%

Supplier: Apollo Scientific

PC9187-1G PC9187-5G
APOSPC9187-1GEA 43.5 EUR
APOSPC9187-1G APOSPC9187-5G
2-Chloro-5-(trifluoromethyl)cinnamic acid 98%
2-Chloro-5-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆ClF₃O₂
MW: 250,602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03002852
CAS Number: 682805-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR