3-(Trifluoromethoxy)phenylacetylene 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Ethynyl-3-(trifluoromethoxy)benzene, 3-Ethynylphenyl trifluoromethyl ether
PC9568-1G
PC9568-5G
PC9568-250MG
APOSPC9568-1GEA
126
EUR
APOSPC9568-1G
APOSPC9568-5G
APOSPC9568-250MG
3-(Trifluoromethoxy)phenylacetylene 98%
3-(Trifluoromethoxy)phenylacetylene
Formula:
C₉H₅F₃O MW: 186,131 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08458148 CAS Number: 866683-57-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...