Perfluorotetraglyme 98%

Supplier: Apollo Scientific

Synonyms: Perfluoro(tetraethylene glycol dimethyl ether)

PC9773-1G PC9773-5G
APOSPC9773-1GEA 94.5 EUR
APOSPC9773-1G APOSPC9773-5G
Perfluorotetraglyme 98%
Perfluorotetraglyme
Formula: C₁₀F₂₂O₅
MW: 618,069 g/mol
Boiling Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD08458173
CAS Number: 64028-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR