2-Bromo-3-methyl-4,4,4-trifluorobutyric acid 97%

Supplier: Apollo Scientific
Danger

PC9848-1G PC9848-5G
APOSPC9848-1GEA 134 EUR
APOSPC9848-1G APOSPC9848-5G
2-Bromo-3-methyl-4,4,4-trifluorobutyric acid 97%
2-Bromo-3-methyl-4,4,4-trifluorobutyric acid
Formula: C₅H₆BrF₃O₂
MW: 234.999 g/mol
Boiling Pt: 92 °C (4 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08458181
CAS Number: 914635-99-7
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR