2-[3-Fluoro-4-(methoxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Apollo Scientific

PC99027-1G PC99027-500MG
APOSPC99027-1GEA 416 EUR
APOSPC99027-1G APOSPC99027-500MG
2-[3-Fluoro-4-(methoxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-[3-Fluoro-4-(methoxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₄H₂₀BFO₃
MW: 266,116 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18732629
CAS Number: 2246867-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR