2-Methyl-4-(trifluoromethoxy)acetanilide

Supplier: Apollo Scientific
Warning

PC9904-1G PC9904-5G
APOSPC9904-1GEA 140 EUR
APOSPC9904-1G APOSPC9904-5G
2-Methyl-4-(trifluoromethoxy)acetanilide
2-Methyl-4-(trifluoromethoxy)acetanilide
Formula: C₁₀H₁₀F₃NO₂
MW: 233,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368678
CAS Number: 886762-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR