2-Fluoro-3-methylbenzyl alcohol 98%
Supplier: Apollo Scientific
|
Warning
|
PC9907-1G
PC9907-5G
APOSPC9907-1GEA
58.2
EUR
APOSPC9907-1G
APOSPC9907-5G
2-Fluoro-3-methylbenzyl alcohol 98%
2-Fluoro-3-methylbenzyl alcohol
Formula:
C₈H₉FO MW: 140,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368780 CAS Number: 307975-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...