Propyl-3,4,5-trihydroxybenzoate pure Ph. Eur., BP, pharma grade
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Gallic acid propyl ester, Propyl gallate, Propyl 3,4,5-trihydroxybenzoate, Progalline P, N-Propyl gallate
BI141962-1KG
BI141962-100G
APOSBI141962-1KGEA
444
EUR
APOSBI141962-1KG
APOSBI141962-100G
Propyl-3,4,5-trihydroxybenzoate pure Ph. Eur., BP, pharma grade
Propyl 3,4,5-trihydroxybenzoate
Formula:
C₁₀H₁₂O₅ MW: 212,2 g/mol Melting Pt: 146…149 °C Density: 0,5 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002196 CAS Number: 121-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...