Mycophenolate mofetil
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Morpholinoethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoate, 2-(4-Morpholinyl)ethyl (E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoate, (E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic Acid 2-(4-Morpholinyl)ethyl Ester
BI1536-250MG
BI1536-500MG
BI1536-100MG
APOSBI1536-250MGEA
212
EUR
APOSBI1536-250MG
APOSBI1536-500MG
APOSBI1536-100MG
Mycophenolate mofetil
Mycophenolate mofetil
Formula:
C₂₃H₃₁NO₇ MW: 433,50 g/mol Melting Pt: 94…95 °C Storage Temperature: Freezer |
MDL Number:
MFCD00867568 CAS Number: 128794-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...