Bleomycin sulphate (mixture)

Supplier: Apollo Scientific
Danger

BI3543-25MG BI3543-5MG
APOSBI3543-25MGEA 386 EUR
APOSBI3543-25MG APOSBI3543-5MG
Bleomycin sulphate (mixture)
Bleomycin sulphate (mixture)

The sulphates of bleomycin are a mixture of basic antineoplastic glycopeptide antibiotics produced by Streptomces verticullus. bleomycin binds to DNA and causes strand scissions.

Formula: C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄
MW: 1512.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070310
CAS Number: 9041-93-4
EINECS: 232-925-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR