Cilostazol

Supplier: Apollo Scientific
Warning

Synonyms: 6-(4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-3,4-dihydro-2(1H)-quinolinone

BI7396-1G BI7396-5G BI7396-25G
APOSBI7396-1GEA 21.9 EUR
APOSBI7396-1G APOSBI7396-5G APOSBI7396-25G
Cilostazol
Cilostazol
Formula: C₂₀H₂₇N₅O₂
MW: 369,47 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00866780
CAS Number: 73963-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR