Diethyl pyrocarbonate BioChemica

Supplier: Apollo Scientific
Warning

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

BIA0881-20ML BIA0881-100ML
APOSBIA0881-20MLEA 118 EUR
APOSBIA0881-20ML APOSBIA0881-100ML
Diethyl pyrocarbonate BioChemica
Diethyl pyrocarbonate

  • Assay (NT): min. 97 %

Formula: O(COOC₂H₅)₂
MW: 162,14 g/mol
Boiling Pt: 160…163 °C
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR