4-Aminoantipyrine

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

BIA1342-500G
APOSBIA1342-500GEA 157 EUR
APOSBIA1342-500G
4-Aminoantipyrine
4-Aminoantipyrine

Used in the determination of glucose in the presenceof phenol and peroxidase.

Formula: C₁₁H₁₃N₃O
MW: 203,24 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR