L-α-Aminoadipic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid
BIA4025-1G
BIA4025-5G
APOSBIA4025-1GEA
34.8
EUR
APOSBIA4025-1G
APOSBIA4025-5G
L-α-Aminoadipic acid
L-α-Aminoadipic acid
Formula:
C₆H₁₁NO₄ MW: 161,16 g/mol Melting Pt: 203…205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002636 CAS Number: 1118-90-7 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...