L-Homocysteine

Supplier: Apollo Scientific
Warning

Synonyms: (S)-2-Amino-4-mercaptobutanoic acid, H-HoCys-OH

BIB6065-100MG BIB6065-1G BIB6065-500MG
APOSBIB6065-100MGEA 109 EUR
APOSBIB6065-100MG APOSBIB6065-1G APOSBIB6065-500MG
L-Homocysteine
L-Homocysteine
Formula: C₄H₉NO₂S
MW: 135,19 g/mol
Melting Pt: 263…265 °C
Storage Temperature: Freezer
MDL Number: MFCD00151320
CAS Number: 6027-13-0
EINECS: 227-891-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR