5-Hydroxy-2-methylindole

Supplier: Apollo Scientific
Warning

BIB6067-1G BIB6067-5G
APOSBIB6067-1GEA 21.9 EUR
APOSBIB6067-1G APOSBIB6067-5G
5-Hydroxy-2-methylindole
5-Hydroxy-2-methylindole
Formula: C₉H₉NO
MW: 147,177 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Freezer
MDL Number: MFCD00022714
CAS Number: 13314-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR