Indole-3-pyruvic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
alpha-Oxo-1H-indole-3-propanoic acid, 1H-Indole-3-propanoic acid, .alpha.-oxo-, 3-(1H-Indol-3-yl)-2-oxopropanoic acid, 3-(3-Indolyl)-2-oxopropanoic acid, 3-Indolylpyruvic acid
BIB6079-1G
BIB6079-5G
BIB6079-2.5G
APOSBIB6079-1GEA
129
EUR
APOSBIB6079-1G
APOSBIB6079-5G
APOSBIB6079-2.5G
Indole-3-pyruvic acid
Indole-3-pyruvic acid
Formula:
C₁₁H₉NO₃ MW: 203,197 g/mol Melting Pt: 215 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00005640 CAS Number: 392-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...