Miltefosine
Supplier: Apollo Scientific
|
Warning
|
BIB6120-1G
BIB6120-2.5G
BIB6120-250MG
APOSBIB6120-1GEA
139
EUR
APOSBIB6120-1G
APOSBIB6120-2.5G
APOSBIB6120-250MG
Miltefosine
Miltefosine
Formula:
C₂₁H₄₆NO₄P Storage Temperature: Ambient |
MDL Number:
MFCD00133396 CAS Number: 58066-85-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...