8-Azido-3,6-dioxaoctanoic acid Cyclohexylamine salt ≥98%

Supplier: Apollo Scientific

Synonyms: Azido-PEG2-acetic acid CHA salt, 2-[2-(2-Azidoethoxy)ethoxy]acetic acid Cyclohexylamine salt

BIBP1063-1G BIBP1063-5G BIBP1063-250MG
APOSBIBP1063-1GEA 219 EUR
APOSBIBP1063-1G APOSBIBP1063-5G APOSBIBP1063-250MG
8-Azido-3,6-dioxaoctanoic acid Cyclohexylamine salt ≥98%
8-Azido-3,6-dioxaoctanoic acid Cyclohexylamine salt
Formula: C₆H₁₁N₃O₄·C₆H₁₃N
MW: 288,35 g/mol
Storage Temperature: Ambient
CAS Number: 2098500-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR