Fmoc-L-Lys(N3)-OH ≥98%
Supplier: Apollo Scientific
Synonyms:
6-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-norleucine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-azidohexanoic acid, Fmoc-ε-azido-Nle-OH, Fmoc-Lys(N₃)-OH, 6-Azido-N-Fmoc-L-norleucine
BIBP1082-250MG
BIBP1082-1G
BIBP1082-5G
APOSBIBP1082-250MGEA
123
EUR
APOSBIBP1082-250MG
APOSBIBP1082-1G
APOSBIBP1082-5G
Fmoc-L-Lys(N3)-OH ≥98%
Fmoc-L-Lys(N3)-OH
Formula:
C₂₁H₂₂N₄O₄ MW: 394,431 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13182319 CAS Number: 159610-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...