Curvularin

Supplier: Apollo Scientific

BIBR1117-1MG BIBR1117-5MG
APOSBIBR1117-1MGEA 94.4 EUR
APOSBIBR1117-1MG APOSBIBR1117-5MG
Curvularin
Curvularin
Storage Temperature: Ambient
CAS Number: 10140-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR