Coelenterazine

Supplier: Apollo Scientific
Warning

Synonyms: 8-Benzyl-2-(4-hydroxybenzyl)-6-(4-hydroxyphenyl)imidazo[1,2-a]pyrazin-3(7H)-one, CLZN, 2-[(4-Hydroxyphenyl)methyl]-6-(4-hydroxyphenyl)-8-(phenylmethyl)-imidazo [1,2-a] pyrazin-3-(7H)-one

BIC1018-1MG BIC1018-5MG
APOSBIC1018-1MGEA 139 EUR
APOSBIC1018-1MG APOSBIC1018-5MG
Coelenterazine
Coelenterazine
Formula: C₂₆H₂₁N₃O₃
MW: 423,472 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467176
CAS Number: 55779-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR