(S)-(+)-Camptothecin

Supplier: Apollo Scientific
Danger

Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

BIC8442-10G BIC8442-1G BIC8442-5G
APOSBIC8442-10GEA 174 EUR
APOSBIC8442-10G APOSBIC8442-1G APOSBIC8442-5G
(S)-(+)-Camptothecin
(S)-(+)-Camptothecin

A potent antitumor antibiotic.

Formula: C₂₀H₁₆N₂O₄
MW: 348,36 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00081076
CAS Number: 7689-03-4
UN: 1544
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR