4-Methylphenyl-2,3,4,6-tetra-O-acetyl-β-D-thiogalactopyranoside 99% min

Supplier: Apollo Scientific

Synonyms: 4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-beta-D-galactopyranoside

BICL4166-10G BICL4166-5G BICL4166-2G
APOSBICL4166-10GEA 978 EUR
APOSBICL4166-10G APOSBICL4166-5G APOSBICL4166-2G
4-Methylphenyl-2,3,4,6-tetra-O-acetyl-β-D-thiogalactopyranoside 99% min
4-Methylphenyl-2,3,4,6-tetra-O-acetyl-beta-D-thiogalactopyranoside
Formula: C₂₁H₂₆O₉S
Storage Temperature: Ambient
MDL Number: MFCD09750783
CAS Number: 28244-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR