Methyl-2,3-O-isopropylidene-β-D-ribofuranoside 98% min
Supplier: Apollo Scientific
Synonyms:
((3aR,4R,6R,6aR)-6-Methoxy-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol
BICL4175-10G
BICL4175-25G
BICL4175-50G
APOSBICL4175-10GEA
253
EUR
APOSBICL4175-10G
APOSBICL4175-25G
APOSBICL4175-50G
Methyl-2,3-O-isopropylidene-β-D-ribofuranoside 98% min
Methyl-2,3-O-isopropylidene-beta-D-ribofuranoside
Formula:
C₉H₁₆O₅ MW: 204,222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00270040 CAS Number: 4099-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...