2-Chloro-4-nitrophenyl-β-D-cellobioside 98% min
Supplier: Apollo Scientific
BICL4224-10MG
BICL4224-25MG
BICL4224-50MG
APOSBICL4224-10MGEA
238
EUR
APOSBICL4224-10MG
APOSBICL4224-25MG
APOSBICL4224-50MG
2-Chloro-4-nitrophenyl-β-D-cellobioside 98% min
2-Chloro-4-nitrophenyl-β-D-cellobioside
Formula:
C₁₈H₂₄ClNO₁₃ Storage Temperature: Ambient |
CAS Number:
135743-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...