Destruxin A (from Metarhizium anisopliae)
Supplier: Apollo Scientific
|
Warning
|
BID1011-1MG
BID1011-100UG
APOSBID1011-1MGEA
696
EUR
APOSBID1011-1MG
APOSBID1011-100UG
Destruxin A (from Metarhizium anisopliae)
Destruxin A
Formula:
C₂₉H₄₇N₅O₇ MW: 577,71 g/mol Storage Temperature: Freezer |
CAS Number:
6686-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...