Destruxin B (from Metarhizium anisopliae)
Supplier: Apollo Scientific
BID1012-1MG
BID1012-100UG
APOSBID1012-1MGEA
696
EUR
APOSBID1012-1MG
APOSBID1012-100UG
Destruxin B (from Metarhizium anisopliae)
Destruxin B
Formula:
C₃₀H₅₁N₅O₇ MW: 593,80 g/mol Melting Pt: 230…240 °C Storage Temperature: Ambient |
CAS Number:
2503-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...