Docetaxel ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: Taxotere

BID2710-100MG BID2710-1G
APOSBID2710-100MGEA 87 EUR
APOSBID2710-100MG APOSBID2710-1G
Docetaxel ≥99%
Docetaxel
Formula: C₄₃H₅₃NO₁₄
MW: 807.89 g/mol
Melting Pt: 186…192 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00800737
CAS Number: 114977-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR