Conessine

Supplier: Apollo Scientific
Warning

BIDF1033-100MG BIDF1033-250MG BIDF1033-25MG
APOSBIDF1033-100MGEA 154 EUR
APOSBIDF1033-100MG APOSBIDF1033-250MG APOSBIDF1033-25MG
Conessine
Conessine

Natural product.

Formula: C₂₄H₄₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD00016752
CAS Number: 546-06-5
EINECS: 208-897-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR