Piperine 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2E,4E)-5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one, 1-[5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoyl]piperidine, 1-Piperoylpiperidine, 1-Piperylpiperidine
BIDF1038-25G
BIDF1038-5G
APOSBIDF1038-25GEA
142
EUR
APOSBIDF1038-25G
APOSBIDF1038-5G
Piperine 96%
Piperine
Natural product.
Formula:
C₁₇H₁₉NO₃ MW: 285,34 g/mol Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005839 CAS Number: 94-62-2 EINECS: 202-348-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...