(+/-)-Equol ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(4-Hydroxyphenyl)chroman-7-ol, (R,S)-Equol, 4′,7-Dihydroxyisoflavane, 7-Hydroxy-3-(4-hydroxyphenyl)chroman, (±)-Equol, 4',7-Isoflavandiol, 3-(4-Hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol

BIE531-1G BIE531-5G BIE531-25G
APOSBIE531-1GEA 53.9 EUR
APOSBIE531-1G APOSBIE531-5G APOSBIE531-25G
(+/-)-Equol ≥99%
(+/-)-Equol

Urinary metabolite of daidzein, produced by humans and horses.

Formula: C₁₅H₁₄O₃
MW: 242,27 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00016662
CAS Number: 94105-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR