Enniatin A

Supplier: Apollo Scientific

BIE9702-1MG
APOSBIE9702-1MGEA 522 EUR
APOSBIE9702-1MG
Enniatin A
Enniatin A
Formula: C₃₆H₆₃N₃O₉
Storage Temperature: Ambient
CAS Number: 2503-13-1
UN: 3462
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR