Topotecan hydrochloride

Supplier: Apollo Scientific

Synonyms: SKF-104864A hydrochloride, 10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride, NSC 609669 hydrochloride, Topotecan HCl, 9-[(dimethylamino)methyl]-10-hydroxy-(20S)-camptothecin hydrochloride, (S)-10-((Dimethylamino)methyl)-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride

BIFK0005-10MG BIFK0005-50MG BIFK0005-100MG
APOSBIFK0005-10MGEA 65.5 EUR
APOSBIFK0005-10MG APOSBIFK0005-50MG APOSBIFK0005-100MG
Topotecan hydrochloride
Topotecan hydrochloride
Formula: C₂₃H₂₃N₃O₅.HCl
MW: 457,91 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866235
CAS Number: 119413-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR