Erlotinib free base ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: 4-[(3-Ethynylphenyl)amino]-6,7-bis(2-methoxyethoxy)quinazoline

BIFK0019-1G BIFK0019-500MG BIFK0019-5G
APOSBIFK0019-1GEA 176 EUR
APOSBIFK0019-1G APOSBIFK0019-500MG APOSBIFK0019-5G
Erlotinib free base ≥99%
Erlotinib

Erlotinib (as the hydrochloride salt) is the active ingredient in the drug product sold under the trade name Tarceva®.

Formula: C₂₂H₂₃N₃O₄
MW: 393,4357 g/mol
Melting Pt: 167…180 °C
Storage Temperature: Freezer
MDL Number: MFCD08063588
CAS Number: 183321-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR