SB 225002 ≥99%
Supplier: Apollo Scientific
Synonyms:
SB225002, 1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea
BIFK0040-10MG
BIFK0040-25MG
BIFK0040-100MG
APOSBIFK0040-10MGEA
65.4
EUR
APOSBIFK0040-10MG
APOSBIFK0040-25MG
APOSBIFK0040-100MG
SB 225002 ≥99%
SB 225002
Formula:
C₁₃H₁₀BrN₃O₄ MW: 352.10 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00954637 CAS Number: 182498-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...