4-Hydroxyphenylretinamide ≥99%

Supplier: Apollo Scientific
Danger

Synonyms: N-(4-Hydroxyphenyl)-all- trans retinamide, Retinoic Acid p-Hydroxyanilide, (2E,4E,6E,8E)-N-(4-Hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenamide, all-trans-N-(4-Hydroxyphenyl)retinamide, Fenretinide, 4-HPR

BIFK0053-100MG BIFK0053-10MG BIFK0053-50MG
APOSBIFK0053-100MGEA 232 EUR
APOSBIFK0053-100MG APOSBIFK0053-10MG APOSBIFK0053-50MG
4-Hydroxyphenylretinamide ≥99%
4-Hydroxyphenylretinamide
Formula: C₂₆H₃₃NO₂
MW: 391.56 g/mol
Melting Pt: 172 °C
Storage Temperature: Freezer
MDL Number: MFCD00792674
CAS Number: 65646-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR