Isoliquiritigenin ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one, 2',4,4'-Trihydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one

BIFK0057-100MG BIFK0057-10MG BIFK0057-250MG
APOSBIFK0057-100MGEA 139 EUR
APOSBIFK0057-100MG APOSBIFK0057-10MG APOSBIFK0057-250MG
Isoliquiritigenin ≥99%
Isoliquiritigenin

Isoliquiritigenin is a licorice chalconoid, a type of natural phenol. It is a soluble guanylyl cyclase activator that possesses antitumor activity.


  • Protein

Formula: C₁₅H₁₂O₄
MW: 256,26 g/mol
Melting Pt: 207…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075907
CAS Number: 961-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR