α-D(+)-Talose

Supplier: Apollo Scientific

BIFS0116-1G BIFS0116-100MG
APOSBIFS0116-1GEA 571 EUR
APOSBIFS0116-1G APOSBIFS0116-100MG
α-D(+)-Talose
α-D-Talose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 124…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00064906
CAS Number: 2595-98-4
EINECS: 219-996-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR