Glycerol monostearate, purified
Supplier: Apollo Scientific
Synonyms:
Glyceryl monostearate
BIG1007-1KG
BIG1007-5KG
APOSBIG1007-1KGEA
139
EUR
APOSBIG1007-1KG
APOSBIG1007-5KG
Glycerol monostearate, purified
Glycerol monostearate
Formula:
C₂₁H₄₄O₅ MW: 376,57 g/mol Melting Pt: 78 °C Storage Temperature: Freezer |
MDL Number:
MFCD00036186 CAS Number: 31566-31-1 EINECS: 250-705-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...