Cefotaxime sodium salt 100 mg/ml

Supplier: Apollo Scientific
Danger

Synonyms: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

BIG1027-10ML BIG1027-100ML
APOSBIG1027-10MLEA 56.8 EUR
APOSBIG1027-10ML APOSBIG1027-100ML
Cefotaxime sodium salt 100 mg/ml
Cefotaxime sodium salt

  • Antibodies

Formula: C₁₆H₁₇N₅O₇S₂
MW: 477,44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079073
CAS Number: 64485-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR