(2-Hydroxylpropyl)-β-cyclodextrine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
BIH6067-100G
BIH6067-25G
APOSBIH6067-100GEA
423
EUR
APOSBIH6067-100G
APOSBIH6067-25G
(2-Hydroxylpropyl)-β-cyclodextrine
(2-Hydroxylpropyl)-β-cyclodextrine
Formula:
C₆₃H₁₁₂O₄₂ Melting Pt: 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD16621721 CAS Number: 128446-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...