Kojic acid 99%

Supplier: Apollo Scientific
Warning

BIK0452-1KG BIK0452-100G BIK0452-500G
APOSBIK0452-1KGEA 132 EUR
APOSBIK0452-1KG APOSBIK0452-100G APOSBIK0452-500G
Kojic acid 99%
Kojic acid
Formula: C₆H₆O₄
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006580
CAS Number: 501-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR