Lincomycin hydrochloride 90%

Supplier: Apollo Scientific
Warning

BIL0127-25G BIL0127-5G BIL0127-100G
APOSBIL0127-25GEA 79.9 EUR
APOSBIL0127-25G APOSBIL0127-5G APOSBIL0127-100G
Lincomycin hydrochloride 90%
Lincomycin hydrochloride

A lincosamide with a primarily bacteriostatic action against gram-negative bacteria


  • Cell culture tested

Formula: C₁₈H₃₄N₂O₆S·HCl
MW: 443.0 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00058237
CAS Number: 859-18-7
EINECS: 212-726-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR