Ophiobolin B

Supplier: Apollo Scientific
Warning

BIM0138-1MG BIM0138-5MG
APOSBIM0138-1MGEA 241 EUR
APOSBIM0138-1MG APOSBIM0138-5MG
Ophiobolin B
Ophiobolin B
Formula: C₂₅H₃₈O₄
Melting Pt: 212 °C
Density: 1,12 g/cm³ (25 °C)
Storage Temperature: Freezer
CAS Number: 5601-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR