2-Iodosobenzoic acid
Supplier: Apollo Scientific
|
Warning
|
BIM111-1G
BIM111-5G
APOSBIM111-1GEA
40.6
EUR
APOSBIM111-1G
APOSBIM111-5G
2-Iodosobenzoic acid
2-Iodosobenzoic acid
Cleaves at carboxyl side of tryptophanRef: Mahoney,W.C.&Hermodson,M.A(1979)Biochemistry 18, 3810-3814
Formula:
C₇H₅IO₃ MW: 264.02 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00002401 CAS Number: 304-91-6 EINECS: 206-159-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...