Salmon GlcA CHA salt, salmon-Gluc CHA salt ≥98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chloro-3-indolyl-beta-D-glucuronic acid cyclohexylammonium salt

BIMB1027-100MG BIMB1027-1G BIMB1027-500MG
APOSBIMB1027-100MGEA 50.9 EUR
APOSBIMB1027-100MG APOSBIMB1027-1G APOSBIMB1027-500MG
Salmon GlcA CHA salt, salmon-Gluc CHA salt ≥98%
Salmon GlcA CHA salt, salmon-Gluc CHA salt

Salmon-XGlcA is an alternative substrate for b-Glucuronidase (GUS) encoded by the gusA gene. In conjunction with X-Gal, useful for simultaneous detection of Gus and Lac activities on the same plate.

Formula: C₁₄H₁₄ClNO₇·C₆H₁₃N
MW: 442,9 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153940
CAS Number: 138182-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR